STIRRED BATH/CIRCULATOR, OPTIMA TXF200 PROGRAMMABLE IMMERSION THERMOSTAT + ST38 STAINLESS STEEL TANK, 38L CAPACITY, INCL DRAIN TAP A ND PUMP. TEMP RANGE AMBIENT +5 TO 200C. 1 * 1 items
Catalog # GRANTXF200-ST38EU
Supplier: GRANT INSTRUMENTS
Frequently Bought Together