ContactUsLinkComponent
STIRRED BATH, OPTIMA T100 IMMERSION THERMOSTAT + ST12 STAINLESS STEEL TANK, 12L CAPACITY, INCL DRAIN TAP. TEMP RANGE AMBIENT +5 TO 1 00C. 1 * 1 items
Catalog # GRANT100-ST12EU
Supplier:  GRANT INSTRUMENTS
STIRRED BATH, OPTIMA T100 IMMERSION THERMOSTAT + ST12 STAINLESS STEEL TANK, 12L CAPACITY, INCL DRAIN TAP. TEMP RANGE AMBIENT +5 TO 1 00C. 1 * 1 items
Catalog # GRANT100-ST12EU
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together