ContactUsLinkComponent
STIRRED BATH/CIRCULATOR, OPTIMA TC120 IMMERSION THERMOSTAT + ST12 STAINLESS STEEL TANK, 12L CAPACITY, INCL. DRAIN TAP AND PUMP. TEMP RANGE AMBIENT +5 TO 120C. 1 * 1 items
Catalog # GRANTC120-ST12EU
Supplier:  GRANT INSTRUMENTS
STIRRED BATH/CIRCULATOR, OPTIMA TC120 IMMERSION THERMOSTAT + ST12 STAINLESS STEEL TANK, 12L CAPACITY, INCL. DRAIN TAP AND PUMP. TEMP RANGE AMBIENT +5 TO 120C. 1 * 1 items
Catalog # GRANTC120-ST12EU
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together