STIRRED BATH/CIRCULATOR, OPTIMA TC120 IMMERSION THERMOSTAT + ST26 STAINLESS STEEL TANK, 26L CAPACITY, INCL. DRAIN TAP AND PUMP. TEMP RANGE AMBIENT+5 TO 120C. INCL. INTEGRATED PUMP. 1 * 1 items
Catalog # GRANTC120-ST26EU
Supplier: GRANT INSTRUMENTS
Frequently Bought Together