Orders LinkContactUsLinkComponent
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
24/29 NS, GL 25, 9 - 11 mm
Catalog # LENZ01302524
Supplier:   Lenz Laborglas
Adapters, glass, cone/screwthread
Catalog # LENZ01302524
Supplier:  Lenz Laborglas
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
24/29 NS, GL 25, 9 - 11 mm
Price
22,40 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread adapters, with screw-cap with aperture and silicone sealing ring with PTFE washers.
Documents
Specifications
  • Cone size:
    24/29 NS
  • Thread:
    GL 25
  • Tube ext.Ø:
    9 - 11 mm
  • Seq:
    09