Orders LinkContactUsLinkComponent
Funnels, powder
Funnels, powder
100 mm, 35 mm, 15 mm
Catalog # KAVA632413102100
Supplier:   KAVALIER GLASS
Funnels, powder
Catalog # KAVA632413102100
Supplier:  KAVALIER GLASS
Funnels, powder
Funnels, powder
100 mm, 35 mm, 15 mm
Price
63,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These funnels with short stem and bevelled edges are suitable for transferring powdery materials.
Ordering information: Ø = internal stem diameter.
Specifications
  • Ø:
    15 mm
  • Funnel O-Ø:
    100 mm
  • Stem length:
    35 mm