Cistern 640 L - ONU certified, U.N. approved, HDPE white, dimensions 1200 x 800 x 1000 mm. Stopcock : Butterfly valve DN50 integrated. Cage: galvanized welded steel pipes. Stainless steel pallet. 1 * 1 items
Catalog # LAMA3204-S
Supplier: LAMAPLAST
Frequently Bought Together