Box, storage, for 15ml cell culture tubes, 5x5 places for 15ml tubes with diameter 17mm, material PP, with two inserts to hold, colo ur pink, neolab. 1 * 1 items
Catalog # KOEH290121919
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Frequently Bought Together