ContactUsLinkComponent
Volumetric pipettes with bulb - amber coloured - with one mark at 1 ml, class A, tolerance indicated, tol. 0,008 ml, conformity certified,acc. to ISO 648 - AS - 20C, Ex 1 * 1 items
  HECH92255603050
undefined
Volumetric pipettes with bulb - amber coloured - with one mark at 1 ml, class A, tolerance indicated, tol. 0,008 ml, conformity certified,acc. to ISO 648 - AS - 20C, Ex 1 * 1 items
  HECH92255603050
Frequently Bought Together