Orders LinkContactUsLinkComponent
Write-on labels
Write-on labels
Write-on labels, blank, WOB-1138-G.M.R, Vinyl, Black on yellow, 38×11 mm, 80 labels/sheet
Catalog # BRDY101808
Supplier:   Brady
Write-on labels
Catalog # BRDY101808
Supplier:  Brady
Write-on labels
Write-on labels
Write-on labels, blank, WOB-1138-G.M.R, Vinyl, Black on yellow, 38×11 mm, 80 labels/sheet
Price
214,00 €
Log in for availability
1 * 25 items
Product Details & Documents
These write-on labels provide easy identification for calibration, testing, and repair of tools and equipment.
  • Labels are used to track calibration, inspection, or parts and/or equipement
Write-on areas allow date, inspector, or other information to be periodically added.
Specifications
  • Colour:
    Black on yellow
  • Description:
    Write-on labels, blank
  • Dimensions:
    38×11 mm
  • Material:
    Vinyl
  • Seq:
    02
  • Model:
    WOB-1138-G.M.R
  • Packed:
    80 labels/sheet