Pipette tips
Extended length (91 mm), 1 - 200 µl, -, Transparent, Pipetman, Eppendorf, Nichiryo, Oxford, Costar, Volac, Rainin, Finnpipette, Bulk
DELT200026
: DELTALAB
- Packed:Bulk
- Sterile:-
- Volume:1 - 200 µl
- Seq:06
- Tip style:Extended length (91 mm)
- Tip colour:Transparent
- For use with:Pipetman, Eppendorf, Nichiryo, Oxford, Costar, Volac, Rainin, Finnpipette