Orders LinkContactUsLinkComponent
Pipette tips
Gilson type, graduated, bevelled, 2 - 200 µl, -, Transparent, Gilson P20/P50/P100/P200, Eppendorf, Socorex, Nichiryo, HTL, Bulk (25×1000)
Catalog # DELT200078
Supplier:   DELTALAB
Pipette tips
Catalog # DELT200078
Supplier:  DELTALAB
Pipette tips
Gilson type, graduated, bevelled, 2 - 200 µl, -, Transparent, Gilson P20/P50/P100/P200, Eppendorf, Socorex, Nichiryo, HTL, Bulk (25×1000)
Price
492,00 €
Log in for availability
1 * 25.000 items
Specifications
  • Packed:
    Bulk (25×1000)
  • Sterile:
    -
  • Volume:
    2 - 200 µl
  • Seq:
    08
  • Tip style:
    Gilson type, graduated, bevelled
  • Tip colour:
    Transparent
  • For use with:
    Gilson P20/P50/P100/P200, Eppendorf, Socorex, Nichiryo, HTL