Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
6 COMPOSES AS,CD,MN,PB,SE,ZN DANS HNO3 5% As 1000mg/l Cd 100mg/l Mn 1000mg/l Pb 1000mg/l Se 1000mg/l Zn 100mg/l 1 * 100 mL
Catalog # CMPLA191.1K.5N.L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
6 COMPOSES AS,CD,MN,PB,SE,ZN DANS HNO3 5% As 1000mg/l Cd 100mg/l Mn 1000mg/l Pb 1000mg/l Se 1000mg/l Zn 100mg/l 1 * 100 mL
Catalog # CMPLA191.1K.5N.L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection