ContactUsLinkComponent
Hand Held Refractometer MASTER-10P Alpfa. MASTER-10 series is suitable for particularly low concentrations of Brix 10% or less, such as tomato juice, vegetables, soups, cleaning solutions and cutting oil. The temperature correction function allows ac 1 * 1 items
Catalog # 635-0670
Supplier:  Atago
undefined
Hand Held Refractometer MASTER-10P Alpfa. MASTER-10 series is suitable for particularly low concentrations of Brix 10% or less, such as tomato juice, vegetables, soups, cleaning solutions and cutting oil. The temperature correction function allows ac 1 * 1 items
Catalog # 635-0670
Supplier:  Atago
Frequently Bought Together