Ferroin Indicator (1,10 - Phenanthroline Ferrous Sulphate - Complex Solution) 1 * 100 mL
Catalog # REGEPFS1
Supplier: REAGECON
Ferroin Indicator (1,10 - Phenanthroline Ferrous Sulphate - Complex Solution) 1 * 100 mL
Catalog # REGEPFS1
Supplier: REAGECON
Frequently Bought Together
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, dehydrated (max. 0.01% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals