Orders LinkContactUsLinkComponent
Micro Tubes with Thread
Micro Tubes with Thread
Micro tubes with skirted base, neutral cap, 0,5 ml, 500/case
Catalog # SARS72.730.217
Supplier:   SARSTEDT
Micro Tubes with Thread
Catalog # SARS72.730.217
Supplier:  SARSTEDT
Micro Tubes with Thread
Micro tubes with skirted base, neutral cap, 0,5 ml, 500/case
Price
91,80 €
Log in for availability
1 * 500 items
Product Details & Documents
PP, translucent with PP screw cap.
  • With printed label
  • Withstand centrifugation up to 20000×g
  • Attached, not screwed-on cap
  • Sterile and free of DNA, DNase/RNase, PCR inhibitors, ATP and pyrogens/endotoxins
With or without skirted base.
Specifications
  • Packed:
    500/case
  • Capacity:
    0,5 ml
  • Description:
    Micro tubes with skirted base, neutral cap
  • Seq:
    1