ContactUsLinkComponent
MOSIEVE-AGAROSE MS-1000 25 G separation of small DNAs, 50 - 2000 bp Specialised agarose high-resolution separation of small nucleic acids. Easy handling due to high gel strength. Free of RNases, DNases and enzyme inhibitors. Shipping: RT Storage: RT 1 * 25 g
Catalog # 732-3189
Supplier:  VWR Peqlab
undefined
MOSIEVE-AGAROSE MS-1000 25 G separation of small DNAs, 50 - 2000 bp Specialised agarose high-resolution separation of small nucleic acids. Easy handling due to high gel strength. Free of RNases, DNases and enzyme inhibitors. Shipping: RT Storage: RT 1 * 25 g
Catalog # 732-3189
Supplier:  VWR Peqlab