MOSIEVE-AGAROSE MS-1000 100 G separation of small DNAs, 50 - 2000 bp Specialised agarose high-resolution separation of small nucleic acids. Easy handling due to high gel strength. Free of RNases, DNases and enzyme inhibitors. Shipping: RT Storage: RT 1 * 100 g
Catalog # 732-3190
Supplier: VWR Peqlab
Related Information
35-40XX_e_v1116_m 35-40XX_v1016_m
Datasheet
peqGOLD MoSieve-Agarose MS-1000
Product Literature
Frequently Bought Together