PEQGOLD 'LOW MELT'-AGAROSE 250 G Preparative separations, 0.08 - 20 kbp Specialised agarose with a low melting point ideal for nucleic acid recovery. Suitable for all common in-gel-techniques. Free of RNases, DNases and enzyme inhibitors. Shipping: R 1 * 250 g
Catalog # 732-3185
Supplier: VWR Peqlab
Frequently Bought Together