PEQGOLD 'LOW MELT'-AGAROSE 25 G Preparative separations, 0.08 - 20 kbp Specialised agarose with a low melting point ideal for nucleic acid recovery. Suitable for all common in-gel-techniques. Free of RNases, DNases and enzyme inhibitors. Shipping: RT 1 * 25 g
Catalog # 732-3183
Supplier: VWR Peqlab
Related Information
35-20XX_e_v1116_m 35-20XX_v1016_m
Datasheet
peqGOLD Low Melt-Agarose
Product Literature
Frequently Bought Together