ContactUsLinkComponent
ULTRA LOW RANGE DNA LADDER I 10 - 300 bp, 500 µl (RTU) Ready-to-use in loading buffer. 11 discrete fragments (100 ng DNA/µl). Including 6x Loading Buffer. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3297
Supplier:  VWR Peqlab
ULTRA LOW RANGE DNA LADDER I 10 - 300 bp, 500 µl (RTU) Ready-to-use in loading buffer. 11 discrete fragments (100 ng DNA/µl). Including 6x Loading Buffer. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3297
Supplier:  VWR Peqlab