ContactUsLinkComponent
ULTRA LOW RANGE DNA LADDER II 25 - 700 bp, 5 x 50 µg 10 discrete fragments in 10 mM Tris-HCl and 1 mM EDTA (500 ng DNA/µl). Including 6x Loading Buffer Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3299
Supplier:  VWR Peqlab
undefined
ULTRA LOW RANGE DNA LADDER II 25 - 700 bp, 5 x 50 µg 10 discrete fragments in 10 mM Tris-HCl and 1 mM EDTA (500 ng DNA/µl). Including 6x Loading Buffer Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3299
Supplier:  VWR Peqlab
Frequently Bought Together