HIGH RANGE DNA LADDER 1.5 - 10 kbp, 2 x 500 µl Ready-to-use in Loading Buffer! 9 discrete fragments (42 ng DNA/µl). Designed for evaluation of DNA fragments's size and quantitiy. Including: 6x Sample Loading Buffer Shipment: Blue Ice Storage: -20 °C 1 * 1 items
732-3336
Frequently Bought Together