ContactUsLinkComponent
O'RANGE 50 BP DNA LADDER 50 - 1,000 bp, 500 µl Ready-to-use in Loading Buffer! 20 discrete fragments from ligation of shorter DNA molecules (50 ng DNA/µl). 1.000 bp , 500 bp and 200 bp fragments have a higher DNA amount. Including 6x Loading Buffer w 1 * 1 items
Catalog # 732-3289
Supplier:  VWR Peqlab
undefined
O'RANGE 50 BP DNA LADDER 50 - 1,000 bp, 500 µl Ready-to-use in Loading Buffer! 20 discrete fragments from ligation of shorter DNA molecules (50 ng DNA/µl). 1.000 bp , 500 bp and 200 bp fragments have a higher DNA amount. Including 6x Loading Buffer w 1 * 1 items
Catalog # 732-3289
Supplier:  VWR Peqlab
Related Information
Frequently Bought Together