ContactUsLinkComponent
O'RANGE 500 BP DNA LADDER 500 - 6,000 bp, 500 µl Ready-to-use in Loading Buffer! 12 discrete fragments from ligation of shorter DNA molecules (50 ng DNA/µl). 6,000 bp and 2,000 bp fragments have a higher DNA amount. Including 6x Orange G Loading Buff 1 * 1 items
Catalog # PEQL25-2330
Supplier:  VWR Peqlab
undefined
O'RANGE 500 BP DNA LADDER 500 - 6,000 bp, 500 µl Ready-to-use in Loading Buffer! 12 discrete fragments from ligation of shorter DNA molecules (50 ng DNA/µl). 6,000 bp and 2,000 bp fragments have a higher DNA amount. Including 6x Orange G Loading Buff 1 * 1 items
Catalog # PEQL25-2330
Supplier:  VWR Peqlab