ContactUsLinkComponent
50 BP DNA LADDER ORANGE G 50 - 1,000 bp, 500 µl Ready-to-use in loading buffer with Orange G. 13 discrete fragments (100 ng DNA/µl). Including 6x Loading Buffer with Orange G. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3291
Supplier:  VWR Peqlab
undefined
50 BP DNA LADDER ORANGE G 50 - 1,000 bp, 500 µl Ready-to-use in loading buffer with Orange G. 13 discrete fragments (100 ng DNA/µl). Including 6x Loading Buffer with Orange G. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3291
Supplier:  VWR Peqlab