ContactUsLinkComponent
1 KB DNA LADDER ORANGE G 250 - 10,000 bp, 5 x 500 µl Ready-to-use in loading buffer with Orange G. 14 discrete fragments (100 ng DNA/µl). Including 6x Orange G loading buffer Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3294
Supplier:  VWR Peqlab
undefined
1 KB DNA LADDER ORANGE G 250 - 10,000 bp, 5 x 500 µl Ready-to-use in loading buffer with Orange G. 14 discrete fragments (100 ng DNA/µl). Including 6x Orange G loading buffer Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3294
Supplier:  VWR Peqlab