ContactUsLinkComponent
LOW RANGE RNA LADDER 100 - 1000 b, 5 x 10 µg 7 RNA transcripts in 1 mM EDTA (500 ng RNA/µl) Including 2x loading buffer. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3301
Supplier:  VWR Peqlab
undefined
LOW RANGE RNA LADDER 100 - 1000 b, 5 x 10 µg 7 RNA transcripts in 1 mM EDTA (500 ng RNA/µl) Including 2x loading buffer. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3301
Supplier:  VWR Peqlab
Related Information
Frequently Bought Together