PEQGOLD FUNGAL DNA MINI KIT 5 Preps Isolation of DNA from of up to 0.1 g material. Yield: 20 - 30 µg Kit Components: PerfectBind DNA Columns, Collection Tubes, Lysis Buffer PL1, Lysis Buffer PL2, DNA Binding Buffer, DNA Wash Buffer, Elution Buffer Sh 1 * 1 KIT
Catalog # 732-3051
Supplier: VWR Collection
Frequently Bought Together