PURE PFU DNA POLYMERASE 100 u For high accuracy PCRs. With Enzyme, Reaction Buffer complete, Reaction Buffer incomplete, Diluent Buffer and 25 mM magnesium sulphate. Shipment: Blue Ice Storage: -20 C 1 * 1 items
Catalog # 732-3263
Supplier: VWR Peqlab
Frequently Bought Together