CONTROL TAQ 250 u Allows a quick confirmation of enzyme addition. Enzyme with red dye, Reaction Buffer, Diluent Buffer, Enhancer Solution P and 25 mM magnesium chloride. Optimized for TAE buffer. Not suitable for TBE buffer. Shipment: Blue Ice Storag 1 * 1 items
Catalog # PEQL01-2120
Supplier: VWR Peqlab
Frequently Bought Together