PEQGOLD DNASE I 5.000 units, conc. 1 unit/µl Non-specificly cleaving DNA endonuclease. Incl. 10x reaction buffer and 25 mM EDTA. RNase free Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3277
Supplier: VWR Collection
Frequently Bought Together