ContactUsLinkComponent
PURE TAQ DNA POLYMERASE 100 u DNA polymerase free of contaminating DNA. With Enzyme, Reaction Buffer, Diluent Buffer, Enhancer Solution P and 25 mM magnesium chloride. Shipment: Blue Ice Storage: -20 C 1 * 1 items
Catalog # PEQL01-9110
Supplier:  VWR Peqlab
PURE TAQ DNA POLYMERASE 100 u DNA polymerase free of contaminating DNA. With Enzyme, Reaction Buffer, Diluent Buffer, Enhancer Solution P and 25 mM magnesium chloride. Shipment: Blue Ice Storage: -20 C 1 * 1 items
Catalog # PEQL01-9110
Supplier:  VWR Peqlab
Related Information
Frequently Bought Together