ContactUsLinkComponent
DNA SIZER III 125 - 21,226 bp, 5 x 50 µg Lambda DNA/EcoR I/Hind III, 13 discrete fragments in 10 mM Tris-HCl and 1 mM EDTA (0.5 mg DNA/ml). Including 6x Loading Buffer Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # PEQL25-2070
Supplier:  VWR Peqlab
undefined
DNA SIZER III 125 - 21,226 bp, 5 x 50 µg Lambda DNA/EcoR I/Hind III, 13 discrete fragments in 10 mM Tris-HCl and 1 mM EDTA (0.5 mg DNA/ml). Including 6x Loading Buffer Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # PEQL25-2070
Supplier:  VWR Peqlab