ContactUsLinkComponent
DNTP-MIX LONG RANGE 8 µmol (solution) 200 µl ready-to-use solution (pH 7.5) with dATP, dCTP, dGTP and dTTP in a concentration of 10 mM each. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3181
Supplier:  VWR Collection
DNTP-MIX LONG RANGE 8 µmol (solution) 200 µl ready-to-use solution (pH 7.5) with dATP, dCTP, dGTP and dTTP in a concentration of 10 mM each. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3181
Supplier:  VWR Collection