LOW RANGE RNA LADER 100 - 1000 b, 5 x 40 µl (RTU) Ready-to-use in loading buffer. 7 RNA transcripts (250 ng RNA/µl). Including 2x loading buffer. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3302
Supplier: VWR Peqlab
Frequently Bought Together