ContactUsLinkComponent
HIGH RANGE RNA LADDER 200 - 6000 b, 5 x 10 µg 8 RNA transcripts in 1 mM EDTA (500 ng RNA/µl) Including 2x loading buffer. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3303
Supplier:  VWR Peqlab
undefined
HIGH RANGE RNA LADDER 200 - 6000 b, 5 x 10 µg 8 RNA transcripts in 1 mM EDTA (500 ng RNA/µl) Including 2x loading buffer. Shipment: Blue Ice Storage: -20 °C 1 * 1 items
Catalog # 732-3303
Supplier:  VWR Peqlab