PERFECTBLUE DUAL GEL SYSTEM TWIN M Gel Size: 16 x 14 cm (Improved Version) Footprint: 25 x 20 x 14 cm Sample Flexibility: max. 48 Buffer Volume: 650 ml Complete System with Internal Cooling Core SuperSafe Cover with Attached Leads 4 Glass Plates 4 No 1 * 1 items
Catalog # 700-0613
Supplier: VWR Peqlab
Related Information
Broschüre_PerfectBlue_DE Doppelgelsystem Twin M
Product Literature
PerfectBlue Twin_EN+DE_m
Manual
Frequently Bought Together