ContactUsLinkComponent
PERFECTBLUE DUAL GEL SYSTEM TWIN S Gel Size: 10 x 10 cm (Improved Version) Footprint: 17 x 14 x 13 cm Sample Flexibility: max. 24 Buffer Volume: 250 ml Complete System with Internal Cooling Core SuperSafe Cover with Attached Leads 4 Glass Plates 2 No 1 * 1 items
Catalog # 700-0437
Supplier:  VWR Peqlab
undefined
PERFECTBLUE DUAL GEL SYSTEM TWIN S Gel Size: 10 x 10 cm (Improved Version) Footprint: 17 x 14 x 13 cm Sample Flexibility: max. 24 Buffer Volume: 250 ml Complete System with Internal Cooling Core SuperSafe Cover with Attached Leads 4 Glass Plates 2 No 1 * 1 items
Catalog # 700-0437
Supplier:  VWR Peqlab
Frequently Bought Together