ContactUsLinkComponent
PERFECTBLUE DUAL GEL SYSTEM TWIN EXW S Gel Size: 20 x 10 cm (Improved Version) Footprint: 30 x 14 x 13 cm Sample Flexibility: max. 72 Buffer Volume: 650 ml Complete System with Internal Cooling Core SuperSafe Cover with Attached Leads 4 Glass Plates 1 * 1 items
Catalog # 700-0438
Supplier:  VWR Peqlab
undefined
PERFECTBLUE DUAL GEL SYSTEM TWIN EXW S Gel Size: 20 x 10 cm (Improved Version) Footprint: 30 x 14 x 13 cm Sample Flexibility: max. 72 Buffer Volume: 650 ml Complete System with Internal Cooling Core SuperSafe Cover with Attached Leads 4 Glass Plates 1 * 1 items
Catalog # 700-0438
Supplier:  VWR Peqlab