ContactUsLinkComponent
PERFECTBLUE GEL SYSTEM MINI M Gel Size 9 x 11 cm Footprint: 15 x 9.5 x 22 cm Sample Flexibility: max. 36 Buffer Volume: 600 ml The electrophoresis system includes: Buffer Chamber with Platinum Electrodes SuperSafe Lid with Attached Leads Gasketed U.V 1 * 1 items
Catalog # 700-0434
Supplier:  VWR Peqlab
undefined
PERFECTBLUE GEL SYSTEM MINI M Gel Size 9 x 11 cm Footprint: 15 x 9.5 x 22 cm Sample Flexibility: max. 36 Buffer Volume: 600 ml The electrophoresis system includes: Buffer Chamber with Platinum Electrodes SuperSafe Lid with Attached Leads Gasketed U.V 1 * 1 items
Catalog # 700-0434
Supplier:  VWR Peqlab
Related Information
Frequently Bought Together