Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MICROTUBE THERMO-SHAKER WITH PSC24 BLOCK FOR 24 X 2.0 ML MICROTUBES, TEMPERATURE RANGE AMBIENT +5 TO 100C, SHAKING SPEED 250 TO 1400 RPM 1 * 1 items
Catalog # 444-0729
Supplier:
GRANT INSTRUMENTS
Expand image
MICROTUBE THERMO-SHAKER WITH PSC24 BLOCK FOR 24 X 2.0 ML MICROTUBES, TEMPERATURE RANGE AMBIENT +5 TO 100C, SHAKING SPEED 250 TO 1400 RPM 1 * 1 items
Catalog # 444-0729
Supplier:
GRANT INSTRUMENTS
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals