ContactUsLinkComponent
Shower, combined shower, body shower with pull rod, fixed mounted eye shower, wall mounted, inlet at top and bottom G3/4 , RAL2002. 1 * 1 items
  118-0138
 :  BROEN-LAB
Shower, combined shower, body shower with pull rod, fixed mounted eye shower, wall mounted, inlet at top and bottom G3/4 , RAL2002. 1 * 1 items
  118-0138
 :  BROEN-LAB
Frequently Bought Together