HPLC-Säule Econosphere Amino 250x4,6mm 5µm old no, 70046 old no: 70046 MANUFACTURER,,Grace ENDCAPPING,,no PORE SIZE,,80 Å PARTICLE SIZE,,5.0 µm INNER DIAMETER,,4.6 mm LENGTH,,250 mm PACKING,,(NH2) Amino COLUMN DESCRIPTION,,Econosphere Please note tha 1 * 1 items
Catalog # MAISA70046
Supplier: DR MAISCH
Frequently Bought Together