Orders LinkContactUsLinkComponent
Gram Negative lysis buffer
Gram Negative lysis buffer
Gram negative lysis buffer, 125 mL
Catalog # 786-566
Supplier:   G-Biosciences
Gram Negative lysis buffer
Catalog # 786-566
Supplier:  G-Biosciences
CAS Number:  
Gram Negative lysis buffer
Gram Negative lysis buffer
Gram negative lysis buffer, 125 mL
CAS Number:  
Price
64,10 €
Log in for availability
1 * 125 mL
Product Details & Documents
Gram-negative lysis buffer is an extraction buffer for soluble proteins from Gram-negative bacteria. It is a proprietary improvement on the lysozyme based lysis in combination with various salts and agents, which allows extraction of soluble proteins. This buffer has been tested for use with several widely used bacteria including E.coli strains.
Specifications
Specifications
  • Pk:
    125 mL
  • Description:
    Gram negative lysis buffer
Specifications