Inoculating loops and needles, disposable
Needle, -, Yellow, 10 needles per sleeve, 300 sleeves per case
Catalog # 391-0529
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Flexible polymer.
- Sterilised by irradiation
- Individually wrapped
- Length: 200 mm
Documents
Certificates
Search by Lot NumberSpecifications
- Packed:10 needles per sleeve, 300 sleeves per case
- Capacity:-
- Colour:Yellow
- Description:Needle
- Seq:09