Pipette tips
Extended, 50 - 1200 µl, +, 90 mm, Purple, Refill pack, 10 trays, 96 each
These pipette tips are suitable for many pipettes on the market.
- Natural colour, premium grade PP, autoclavable
- Excellent seal design for accurate, leak-tight sealing
- Produced and packed in a dust-free environment under strictly maintained conditions
Ordering information: Available in bulk packaging, in autoclavable racks and in refills.
Caution: Article 613-4153 (BHIT780310) is designed for use with the Midi Plus Pipette Controller only.
- Colour code:Purple
- Packed:Refill pack, 10 trays, 96 each
- Sterile:+
- Volume:50 - 1200 µl
- Length:90 mm
- Seq:36
- Tip style:Extended
- Cat. No.:613-1794
- Environmental certification:Renewable energy certificate; ISO 14001
- Environmentally Preferable:Low Manufacturing Impact