JB Aqua 2S Plus, 2 litre Shallow Analogue unstirred water bath thermostatically controlled. Temp range: Ambient+5C to 98C. Temp stability at 37C: 1.0C. Mains supply: 230V AC 50/60Hz. Includes polycarbonate lid, base tray and EURO cordset 1 * 1 items
Catalog # 462-0181
Supplier: GRANT INSTRUMENTS
Frequently Bought Together