JB Aqua 26 Plus, 26 litre Analogue unstirred water bath thermostatically controlled. Temp range: Ambient+5C to 98C. Temp stability at 37C: 1.0C. Mains supply: 230V AC 50/60Hz. Includes polycarbonate lid, base tray and UK cordset 1 * 1 items
462-0178
Frequently Bought Together