Stirring-heating plate with 15 stirring positions, universally applicable for miscellaneous vessel sizes and shapes, stirring point distance 65 mm, suitable for 15 x 250 ml glass beakers 1 * 1 items
Catalog # 442-0766
Supplier: 2MAG
Frequently Bought Together